Isopropyl myristate
Supplier: USP
Synonyms:
Isopropyl tetradecanoate
, Isopropyl Myristate
, Tetradecylic acid isopropyl ester
, Myristic acid isopropyl ester
, Tetradecanoic acid isopropyl ester
1350400
101095-266EA
453.85
USD
101095-266
Isopropyl myristate
Isopropyl myristate
Formula:
C₁₇H₃₄O₂ MW: 270.46 g/mol Boiling Pt: 320 °C (1013 hPa) Melting Pt: 0…1 °C Density: 0.852 g/cm³ (20 °C) Flash Pt: 150 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00008982 CAS Number: 110-27-0 EINECS: 203-751-4 Merck Index: 12,05234 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...