Indigo carmine

Supplier: USP
Warning

Synonyms: CI 730155,5'-Indigodisulfonic acid, Disodium saltCarmine BlueFood blue No. 2Sodium 3,3'-dioxo-[2,2'-biindolinylidene]-5,5'-disulphonateSodium 3,3'-dioxo-[2,2'-biindolinylidene]-5,5'-disulfonateacid Blue 745,5'-Indigodisulphonic acid, Disodium saltFD and C Blue No. 2FD&C Blue #2

1339000
101095-204EA 449.23 USD
101095-204
Indigo carmine
Indigo carmine
Formula: C₁₆H₈N₂Na₂O₈S₂
MW: 466.36 g/mol
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00005723
CAS Number: 860-22-0
EINECS: 212-728-8
Merck Index: 13,04969

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR