Hydrocortisone

Supplier: USP

1316004
101095-150EA 435.38 USD
101095-150
Hydrocortisone
Hydrocortisone
Formula: C₂₁H₃₀O₅
MW: 362.47 g/mol
Boiling Pt: 567 °C (1013 hPa)
Melting Pt: 202…204 °C
Density: 1,4 g/cm³ (20 °C)
MDL Number: MFCD00011654
CAS Number: 50-23-7
EINECS: 200-020-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR