Guaiacol

Supplier: USP
Warning

Synonyms: 2-Methoxyphenol2-Hydroxyanisole1-Hydroxy-2-methoxybenzene

1300004
101095-102EA 407.69 USD
101095-102
Guaiacol
Guaiacol
Formula: C₇H₈O₂
MW: 124.14 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: 28…32 °C
Density: 1.1534 g/cm³ (0 °C)
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00002185
CAS Number: 90-05-1
EINECS: 201-964-7
UN: 2810
ADR: 6.1,III
Merck Index: 13,04568

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR