Iron(II) sulfate heptahydrate

Supplier: USP
Warning

Synonyms: Ferrous sulphate heptahydrateIron vitriolFerrous sulfate heptahydrate

1270355
101095-002EA 420 USD
101095-002
Iron(II) sulfate heptahydrate
Iron (II) sulphate heptahydrate
Formula: FeSO₄·7H₂O
MW: 278.02 g/mol
Melting Pt: 64 °C
Density: 1,9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149719
CAS Number: 7782-63-0
EINECS: 231-753-5
Merck Index: 13,04091

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR