Dehydroacetic acid

Supplier: USP
Warning

Synonyms: 3-Acetyl-6-methyl-2H-pyran-2,4(3H)-dione3-Acetyl-4-hydroxy-6-methyl-2H-pyran-2-one

1166309
101094-734EA 453.85 USD
101094-734
Dehydroacetic acid
Dehydracetic acid
Formula: C₈H₈O₄
MW: 168.15 g/mol
Boiling Pt: 333 °C (1013 hPa)
Melting Pt: 108 °C
Flash Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00066709
CAS Number: 520-45-6
EINECS: 208-293-9
Merck Index: 13,02885

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR