L(+)-Cysteine hydrochloride monohydrate

Supplier: USP

Synonyms: (R)-2-Amino-3-mercaptopropanoic acid hydrochloride hydrate(R)-(+)-Cysteine hydrochloride monohydrateL-Cysteine HCI MonohydrateCysteine Hydrochloride monohydrateH-Cys-OH.HCl.H2OCysteine Hydrochloride, Monohydrate

1161509
101094-724EA 444.62 USD
101094-724
L(+)-Cysteine hydrochloride monohydrate
L(+)-Cysteine hydrochloride monohydrate
Formula: HSCH₂CH(NH₂)COOH·HCl·H₂O
MW: 175.64 g/mol
Melting Pt: 168…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00065606
CAS Number: 7048-04-6
EINECS: 200-157-7
Merck Index: 13,02810
https://us.vwr.com/stibo/web/std.lang.all/58/37/2345837.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR