Adenine

Supplier: USP
Danger

Synonyms: 7H-Purin-6-amine6-AminopurinePurin-6-amine

1012101
101094-214EA 378.46 USD
101094-214
Adenine
Adenine
Formula: C₅H₅N₅
MW: 135.13 g/mol
Density: 0.99741 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041790
CAS Number: 73-24-5
EINECS: 200-796-1
UN: 2811
ADR: 6.1,III
Merck Index: 13,00152

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR