Taurine
Supplier: R&D Systems
Synonyms:
2-Aminoethanesulphonic acid
, 2-Aminoethanesulfonic acid
0209/1G
100505-082EA
112.8
USD
100505-082
Taurine
Taurine
Formula:
C₂H₇NO₃S MW: 125.15 g/mol Melting Pt: 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008197 CAS Number: 107-35-7 EINECS: 203-483-8 Merck Index: 12,09241 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...