Zinc selenide, crystals 45 Degree / Parallelogram Spectrophotometry

Supplier: INTERNATIONAL CRYSTAL LABS MS
Danger

Synonyms: Zinc (II) selenide

0003J-3856 0003J-1634
100213-224EA 850.62 USD
100213-224 100213-096
Zinc selenide, crystals 45 Degree / Parallelogram Spectrophotometry
Zinc selenide

Zinc Selenide (ZnSe) is commonly used for CO2 laser focusing lenses, night vision applications, ATR prisms and transmission windows for IR spectroscopy.

Formula: SeZn
MW: 144.35 g/mol
Melting Pt: 1793
Density: 5.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011296
CAS Number: 1315-09-9
EINECS: 215-259-7
UN: 3283
ADR: 6.1 III
Merck Index: 12,10290

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR