Potassium chloride, crystal random cuttings for spectrophotometry

Supplier: INTERNATIONAL CRYSTAL LABS MS

Synonyms: Potassiumchloride

0002-5583 0002-2499 0002-4538 0002-4980
100214-474EA 93.4 USD
100214-474 100215-918 100212-534 100211-360
Potassium chloride, crystal random cuttings for spectrophotometry
Potassium chloride

Samples being analyzed by diffuse reflectance are often combined with a powder so that a matrix can be formed. KBr and KCl are excellent media for diffuse reflection sampling and for pellets.


KBr and KCl sold as random cuttings can be ground finely immediately prior to being mixed with the sample for either diffuse reflection or transmission pellets. Random cuttings ground into powder a short time before being mixed with a sample will contain

Formula: KCl
Boiling Pt: 1420 °C (1013 hPa)
Melting Pt: 773 °C
Density: 1.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR