Zinc carbonate basic ZnO ≥ 65%, powder, Reagent Grade suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Dicarbonatohexahydroxypentazinc, Zinc carbonate hydroxide, Zinc hydroxide carbonate

Z1050-12KG Z1050-500GM Z1050-25GM Z1050-125GM Z1050-2.5KG
700016-304EA 1880.45 USD
700016-304 700016-310 700016-308 700016-302 700016-306
Zinc carbonate basic ZnO ≥ 65%, powder, Reagent Grade suitable for use as excipient
Zinc carbonate basic

BSE/TSE statement (available upon request) Allergen statement (available upon request)


BSE/TSE statement (available upon request)
Allergen statement (available upon request)

Formula: C₂H₆O₁₂Zn₅
MW: 549.01 g/mol
Density: 4.3476 g/cm³ (23.2 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011519
CAS Number: 5263-02-5
EINECS: 222-477-6

Order Now

Specification Test Results

Assay 65% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR