Xanthan gum NF excipient_suitable

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Xanthan

XA105-100GM XA105-500GM XA105-12KG XA105-2.5KG
95036-600EA 126.06 USD
95036-600 95036-604 700000-078 95036-602
Xanthan gum NF excipient_suitable
Xanthan gum

Xanthan Gum, NF is a natural gum polysaccharide used as a thickener or rheology modifier and emulsion stabilizer. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00131256
CAS Number: 11138-66-2
EINECS: 234-394-2

Order Now

Specification Test Results

Assay 91% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR