Uracil ≥98%, Reagent Grade

Supplier: Spectrum Chemical Mfg. Corp.

UR105-100GM UR105-1KG UR105-25GM UR105-500GM
700008-832EA 100.87 USD
700008-832 700008-834 700008-836 700008-838
Uracil ≥98%, Reagent Grade
Uracil

Uracil, Reagent is a naturally occurring pyrimidine derivative and can be used for drug delivery and as a pharmaceutical. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: C₄H₄N₂O₂
Boiling Pt: 379 °C (1013 hPa)
Melting Pt: 194 °C
Density: 0.99744 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006016
CAS Number: 66-22-8
EINECS: 200-621-9
Merck Index: 12,09985

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR