Gum tragacanth, powder NF

Supplier: Spectrum Chemical Mfg. Corp.

TR105-50KGBL TR105-12KG TR105-125GM TR105-2.5KG TR105-500GM
700000-076EA 7511.2 USD
700000-076 700000-074 95036-354 95036-356 95036-358
Gum tragacanth, powder NF
Gum tragacanth

Tragacanth, Powder, NF is used as an emulsifier, thickener, stabilizer, and texturant additive. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Storage Temperature: Ambient
MDL Number: MFCD00131255
CAS Number: 9000-65-1
EINECS: 232-552-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR