Taurine ≥98.5%, Reagent Grade
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
2-Aminoethanesulphonic acid
, 2-Aminoethanesulfonic acid
Taurine, Reagent is an organic acid found in animal tissues, specifically bile and the large intestine. Some of its roles include conjugation of bile acids, membrane stabilizations and modulation of calcium signaling. It is crucial for cardiovascular function. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.
Formula:
C₂H₇NO₃S MW: 125.15 g/mol Melting Pt: 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008197 CAS Number: 107-35-7 EINECS: 203-483-8 Merck Index: 13,09163 |
Specification Test Results
Assay | 98.5% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...