2,2,4-Trimethylpentane 99 ACS for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane

T1125-200LTB T1125-4LTGL T1125-1LTGL T1125-20LT T1125-500MLG
700002-272EA 6103.99 USD
700002-272 700002-276 700002-270 700002-274 700002-278
2,2,4-Trimethylpentane 99 ACS for synthesis
2,2,4-Trimethylpentane
Formula: (CH₃)₂CHCH₂C(CH₃)₃
MW: 114.23 g/mol
Boiling Pt: 99.2
Melting Pt: -107
Density: 0.692 g/cm³ (20 °C)
Flash Pt: –12 °C
Storage Temperature: Ambient
MDL Number: MFCD00008943
CAS Number: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3 II
Merck Index: 13,05212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=bb9d5eb96909b6bebc6a8709db78c4ea0ef52430&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR