Sodium dihydrogen phosphate dihydrate ≥99%
Supplier: Spectrum Chemical Mfg. Corp.
Sodium Phosphate, Monobasic, Dihydrate is one of the sodium salts derived from phosphoric acid. Highly hygroscopic and water soluble it comes in the form of a white powder. In an aqueous solution it is basic, with a pH value between 8.0 and 11.0. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Formula:
NaH₂PO₄·2H₂O Boiling Pt: ∼650 °C (1013 hPa) Melting Pt: 60 °C Density: 1.91 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149209 CAS Number: 13472-35-0 EINECS: 231-449-2 REACH: 01-2119489796-13 Merck Index: 12,08806 |
Specification Test Results
Assay | 99% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...