Sulfanilic acid, anhydrous ≥98% ACS for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Aniline-4-sulphonic acid, 4-Aminobenzenesulphonic acid, 4-Aminobenzenesulfonic acid, Aniline-4-sulfonic acid

S1703-125GM S1703-12KGBL S1703-500GM S1703-50KGBL S1703-25GM
700006-836EA 259.52 USD
700006-836 700006-838 700006-842 700006-844 700006-840
Sulfanilic acid, anhydrous ≥98% ACS for synthesis
Sulphanilic acid

Sulfanilic Acid, Anhydrous, Reagent, ACS, also known as 4-aminobenzenesulfonic acid, is used to make sulpha drugs and dyes, as well as for the quantitative analysis of nitrite and nitrate ions.


As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Formula: 4-(H₂N)C₆H₄SO₃H
MW: 173.19 g/mol
Boiling Pt: 363
Melting Pt: 279
Density: 1.485 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007886
CAS Number: 121-57-3
EINECS: 204-482-5
Merck Index: 13,09012
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e032bed891f24ee48419dc3c7237b0293fd3e629&vid=28238ab814bb2b8d6eb5012387d8bf41528469e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR