Starch (from wheat), powder

Supplier: SPECTRUM CHEMICAL MFG CORP

S1557-2.5KG S1557-500GM S1557-12KG
700011-356EA 240.87 USD
700011-356 700011-358 700011-354
Starch (from wheat), powder
Starch

Starch, Wheat, Powder is a carbohydrate and polysaccharide that has a large number of glucose units that are linked by glycosidic bonds derived from wheat. It is primarily used as a food additive and used in the fabrication of sweets such as wine gums and jelly beans where it is used as a mould. It is also used in the paper industry, clothing industry, as an adhesive and in the construction industry.

Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-25-8
EINECS: 232-679-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR