Starch Potato, powder suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP

S1553-500GM S1553-50KGBL S1553-2.5KG S1553-12KG
700011-344EA 118.71 USD
700011-344 700011-346 700011-342 700011-340
Starch Potato, powder suitable for use as excipient
Starch

Starch, Potato, Powder is a carbohydrate and polysaccharide that has a large number of glucose units that are linked by glycosidic bonds derived from potatoes. It is primarily used as a food additive and used in the fabrication of sweets such as wine gums and jelly beans where it is used as a mould. It is also used in the paper industry, clothing industry, as an adhesive and in the construction industry.

Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-25-8
EINECS: 232-679-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR