Disodium oxalate ≥99.5%, powder ACS for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Sodium ethanedioate, Sodium oxalate, Soerensen's buffer substances

S1370-12KG S1370-500GM S1370-125GM S1370-50KGBL S1370-2.5KG
700005-782EA 3834.2 USD
700005-782 700005-786 700005-780 700005-788 700005-784
Disodium oxalate ≥99.5%, powder ACS for synthesis
di-Sodium oxalate

Sodium Oxalate, Powder, Reagent, ACS is the white crystalline, odorless sodium salt of oxalic acid. Used as a reducing agent, it can be used as a primary standard for potassium permanganate solution standardizing. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness

Formula: Na₂C₂O₄
Melting Pt: 250…270 °C
Density: 2.27 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012465
CAS Number: 62-76-0
EINECS: 200-550-3
Merck Index: 13,08723

Order Now

Specification Test Results

Assay 99.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR