Sodium formate ≥99%, crystals ACS

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Formic acid sodium salt

S1285-500GM S1285-50KGBL S1285-12KG S1285-2.5KG S1285-125GM
700004-228EA 2348.04 USD
700004-228 700004-222 700004-220 700004-226 700004-224
Sodium formate ≥99%, crystals ACS
Sodium formate

Sodium Formate, Crystal, Reagent, ACS is the sodium salt of formic acid and appears as a deliquescent white powder. It is typically used as a buffering agent for mineral acids, as well as food additive.


As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Formula: HCO₂Na
MW: 68.01 g/mol
Melting Pt: 257.31 °C
Density: 1.919 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013101
CAS Number: 141-53-7
EINECS: 205-488-0
REACH: 01-2119486468-21
Merck Index: 12,08765

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR