Sodium chloride ≥99%, granular FCC, non-iodised

Supplier: Spectrum Chemical Mfg. Corp.

S1247-500GM S1247-45KGBL S1247-12KG S1247-2.5KG
95035-764EA 116.63 USD
95035-764 700000-328 700000-326 95035-762
Sodium chloride ≥99%, granular FCC, non-iodised
Sodium chloride

Sodium Chloride, Non-Iodized, Granular, FCC is used as a flavor enhancer and preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: NaCl
Boiling Pt: 1413 °C (1013 hPa)
Melting Pt: 801 °C
Density: 2.16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR