L(+)-Potassium sodium tartrate tetrahydrate ≥99%, powder USP
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Potassium sodium tartrate tetrahydrate, Potassium sodium (2R,3R)-2,3-dihydroxysuccinate tetrahydrate, Potassium sodium L(+)-tartrate tetrahydrate, Sodium potassium tartrate, (R,R)-(+)-Tartaric acid potassium sodium salt tetrahydrate, (R,R)-(+)-Potassium sodium tartrate tetrahydrate, Rochelle salt, L(+)-Tartaric acid potassium sodium salt tetrahydrate, (+)-Potassium sodium L-tartrate tetrahydrate
Form: Powder
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
KOOC(CHOH)₂COONa·4H₂O MW: 282.22 g/mol Melting Pt: 70…80 °C Density: ∼1.767 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00150989 CAS Number: 6381-59-5 EINECS: 206-156-8 Merck Index: 13,07755 |
Specification Test Results
Assay | 99% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...