Phloroglucinol, anhydrous
Supplier: Spectrum Chemical Mfg. Corp.
|
Warning
|
Phloroglucinol, Anhydrous is an organic compound phenol derivative with antispasmodic properties. It is used as a coupling agent, linking diazo dyes to give a fast black. In cosmetic and personal care product formulations, it is used for its colorant and antioxidant properties. It appears as a light yellow powder.
Formula:
C₆H₆O₃ Boiling Pt: 400 °C (1013 hPa) Melting Pt: 217…220 °C Density: 1.46 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002286 CAS Number: 108-73-6 EINECS: 203-611-2 Merck Index: 12,07482 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...