Propylene carbonate, Reagent Grade

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Propylene glycol carbonate, 4-Methyl-1,3-dioxolan-2-one, Carbonic acid propylene glycol ester

P1433-200LTB P1433-25GM P1433-3KG P1433-20KG P1433-500GM
700008-540EA 3007.05 USD
700008-540 700008-544 700008-546 700008-542 700008-548
Propylene carbonate, Reagent Grade
Propylene carbonate

Propylene Carbonate, Reagent, is an organic compound that is a liquid that is both colorless and odorless. Used in a range of applications, the most notable one is as a polar and aprotic solvent in syntheses. It is also used as a high-permittivity component as it has a dielectric constant of 64. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Formula: C₄H₆O₃
MW: 102.09 g/mol
Boiling Pt: 241…242 °C (1013 hPa)
Melting Pt: -48.8
Density: 1.189
Flash Pt: 132 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00005385
CAS Number: 108-32-7
EINECS: 203-572-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR