Potassium hydroxide ≥90%, soluble, Technical Grade for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: Potash caustic, Potassium hydrate, Caustic potash

P1325-500GM P1325-12KG P1325-2.5KG
700000-782EA 106.12 USD
700000-782 700000-778 700000-780
Potassium hydroxide ≥90%, soluble, Technical Grade for synthesis
Potassium hydroxide

Potassium Hydroxide, Flakes, Technical is an inorganic compound commonly referred to as Caustic Potash. A colorless solid chemical compound and is a strong base. Technical grade products supplied by Spectrum are indicative of a grade suitable for general industrial use. These materials may or may not have a Certificate of Analysis available.

Formula: KOH
Boiling Pt: 1320 °C (1013 hPa)
Melting Pt: 360 °C
Density: 2.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1813
ADR: 8,II
REACH: 01-2119487136-33
Merck Index: 12,07806

Order Now

Specification Test Results

Assay 90% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR