Octyl acetate ≥99% for synthesis
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Acetic acid octyl ester
O2081-100GM
O2081-25GM
700004-600EA
122.8
USD
700004-600
700004-602
Octyl acetate ≥99% for synthesis
Octyl acetate
Octyl Acetate, also known as Octyl ethanoate or n-Octyl acetate, is an organic compound used as a solvent for waxes, oils, and nitrocellulose; it is also the primary ingredient for artificial flavors and some perfumes.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₀H₂₀O₂ Boiling Pt: 195…199 °C (1013 hPa) Melting Pt: –38.5 °C Density: 0.867 Storage Temperature: Ambient |
MDL Number:
MFCD00009562 CAS Number: 112-14-1 EINECS: 203-939-6 |
Specification Test Results
Assay | 99% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...