Nicotinamide USP

Supplier: Spectrum Chemical Mfg. Corp.
Warning

NI105-500GM NI105-5KG NI105-1KG NI105-25KGBL NI105-100GM
95034-750EA 151.3 USD
95034-750 700009-446 95034-748 700009-444 95034-746
Nicotinamide USP
Nicotinamide

Niacinamide, USP is a form of vitamin B3. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₆H₆N₂O
Boiling Pt: 334 °C (1013 hPa)
Melting Pt: 127…128 °C
Density: 1.4463 g/cm³ (20 °C)
Flash Pt: 182 °C
Storage Temperature: Ambient
MDL Number: MFCD00006395
CAS Number: 98-92-0
EINECS: 202-713-4
Merck Index: 12,06574

Order Now

Specification Test Results

Assay 98.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR