Methyl salicylate ≥98% NF suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Wintergreen Oil, Methyl 2-hydroxybenzoate

ME170-20LTMT ME170-4LTGL ME170-500MLG ME170-500MLG ME170-200LTB ME170-200LTB
89078-882EA 2059.18 USD
89078-882 95034-448 700006-426 95034-450 700006-424 89078-884
Methyl salicylate ≥98% NF suitable for use as excipient
Methyl salicylate

All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: 2-(HO)C₆H₄CO₂CH₃
Boiling Pt: 432 °C (1013 hPa)
Melting Pt: 100…101 °C
Density: 1.184
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00002214
CAS Number: 119-36-8
EINECS: 204-317-7
Merck Index: 13,06143
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d636eb895c055579d494c4b8ed1636c32f052dd7&vid=7f7da4bc95980831012b381705f4b5a3f4ab7003&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR