D-(-)-Mannitol ≥96%, powder USP suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol

MA165-50KGBL MA165-2.5KG MA165-12KG MA165-500GM MA165-125GM
89050-440EA 3392.55 USD
89050-440 95034-336 89050-438 95034-338 95034-334
D-(-)-Mannitol ≥96%, powder USP suitable for use as excipient
D(-)-Mannitol

Mannitol, Powder, USP. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: HOH₂C(CH(OH))₄CH₂OH
MW: 182.17 g/mol
Boiling Pt: 290…295 °C (4 hPa)
Melting Pt: 164…169 °C
Density: 1.49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064287
CAS Number: 69-65-8
EINECS: 200-711-8
Merck Index: 13,05769
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=163f556dca45086e05ff490ddc8eeb65d04aea05&vid=408e3f5566e309064ce68988cdd160305c5b8c0a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 96% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR