D-(+)-Maltose monohydrate 92, powder, reagent Biochemistry

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Maltobiose hydrate, Malt Sugar, beta-Maltose monohydrate, Maltose monohydrate, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal hydrate, 4-O-α-D-Glucopyranosyl-D-glucose hydrate

MA155-100KGB MA155-500GMP MA155-100GMP MA155-12KG
700000-256EA 7543.5 USD
700000-256 700000-262 700000-260 700000-254 700000-258
D-(+)-Maltose monohydrate 92, powder, reagent Biochemistry
D(+)-Maltose monohydrate

Maltose, Powder, Reagent, is also referred to as maltbiose or malt sugar. Maltose is a disaccharide that occurs naturally in germinating seeds, when the seed breaks down starch to use as nutrient. It can also be obtained from caramelizing glucose. It is often used as a sweetener and preservative as well being used in the production of beer. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Formula: C₁₂H₂₂O₁₁·1H₂O
MW: 360.32 g/mol
Melting Pt: 160…165 °C
Storage Temperature: Ambient
MDL Number: MFCD00149343
CAS Number: 6363-53-7
EINECS: 200-716-5
Merck Index: 13,05736
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f861d7adf53134a0c485b5942d3be34c4d0b571&vid=16e5c4425a7a8e158a4200470a45483c7ff5781e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Maltose) 92% Min.
ASSAY (GLUCOSE) max. 3.0 %
LOSS ON DRYING max. 7.0 %
HEAVY METALS (as Pb) max. 0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR