Magnesium oxide light ≥96%, powder USP suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Magnesia usta

MA120-12KG MA120-500GM MA120-2.5KG
700000-406EA 2333.6 USD
700000-406 95034-232 95034-230
Magnesium oxide light ≥96%, powder USP suitable for use as excipient
Magnesium oxide

BSE/TSE statement (available upon request) Kosher certified


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: MgO
MW: 40.3 g/mol
Boiling Pt: 3600
Melting Pt: 2800
Density: 3.58 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011109
CAS Number: 1309-48-4
EINECS: 215-171-9
Merck Index: 13,05700

Order Now

Specification Test Results

Assay 96% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR