Mercury (II) oxide, yellow ≥99%, powder ACS

Supplier: Spectrum Chemical Mfg. Corp.
Danger

M1180-500GM M1180-125GM M1180-12KG M1180-45KGBL M1180-2.5KG M1180-25GM
700002-158EA 1312.65 USD
700002-158 700002-148 700002-150 700002-156 700002-152 700002-154
Mercury (II) oxide, yellow ≥99%, powder ACS
Mercury (II) oxide

Form: Powder

Formula: HgO
Melting Pt: 500 °C
Storage Temperature: Ambient
MDL Number: MFCD00011045
CAS Number: 21908-53-2
EINECS: 244-654-7
UN: 1641
ADR: 6.1,II
Merck Index: 13,05908

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR