Manganese(II) chloride tetrahydrate ≥98% FCC suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Manganese dichloride tetrahydrate, Manganese chloride (tetrahydrate), Manganese(II) chloride tetrahydrate, Manganous chloride tetrahydrate, Manganese chloride tetrahydrate, Manganese Chloride, Tetrahydrate

M1109-125GM M1109-2.5KG M1109-500GM
95034-292EA 168.11 USD
95034-292 95034-294 95034-296
Manganese(II) chloride tetrahydrate ≥98% FCC suitable for use as excipient
Manganese (II) chloride tetrahydrate

Manganese Chloride, Tetrahydrate, FCC is used as a dietary supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: MnCl₂·4H₂O
MW: 197.9 g/mol
Boiling Pt: 1190
Melting Pt: 58…59 °C
Density: 2.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149792
CAS Number: 13446-34-9
EINECS: 231-869-6
UN: 3077
ADR: 9 III
Merck Index: 13,05751

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR