Lactose, anhydrous NF

Supplier: Spectrum Chemical Mfg. Corp.

LA103-2.5KG LA103-500GM LA103-12KG LA103-45KGBL
95033-796EA 570.48 USD
95033-796 95033-798 700000-242 700000-244
Lactose, anhydrous NF
Lactose

Anhydrous Lactose, NF is used as a tablet diluent in the pharmaceutical industry. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: C₁₂H₂₂O₁₁
Boiling Pt: 592 °C (1013 hPa)
Melting Pt: 222.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00151251
CAS Number: 63-42-3
EINECS: 200-559-2
Merck Index: 12,05356

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR