N-Lauroylsarcosine sodium salt ≥97%, Ultrapure for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: Lauroylsarcosine sodium, Sarkosyl, Sodium lauroyl sarcosine, Sodium N-lauroylsarcosinate, Sodium N-dodecanoyl-N-methylglycinate

L3115-25GM L3115-500GM L3115-100GM
700007-702EA 101.55 USD
700007-702 700007-704 700007-700
N-Lauroylsarcosine sodium salt ≥97%, Ultrapure for synthesis
N-Lauroylsarcosine sodium salt
Formula: H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa
Melting Pt: 45…47 °C
Flash Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00042728
CAS Number: 137-16-6
EINECS: 205-281-5
UN: 2811
ADR: 6.1,II
Merck Index: 12,04379
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0126d80345cce5dd675a83796c379bd44cebacef&vid=48c74917d6a93a32467317ef6c6fdbf24f3b6dc2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 97% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR