2-Oxoglutaric acid ≥99% for biochemistry

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: 2-Ketoglutaric acid, A-Ketoglutaric acid, α-Ketoglutaric acid, 2-Oxopentanedioic acid

KE110-5GM KE110-100GM KE110-1KG KE110-25GM KE110-5KGBL
700006-566EA 101.1 USD
700006-566 700006-560 700006-562 700006-564 700006-568
2-Oxoglutaric acid ≥99% for biochemistry
2-Oxoglutaric acid

alpha-Ketoglutaric Acid is one of the ketone derivatives of glutaric acid and is a biological compound. It is the resulting keto acid that is created by the deamination of glutamate.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: HOOCCH₂CH₂COCOOH
Boiling Pt: 323 °C (1013 hPa)
Melting Pt: 112…116 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004165
CAS Number: 328-50-7
EINECS: 206-330-3
Merck Index: 13,05320

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR