Indigo carmine ACS
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
CI 73015
, 5,5'-Indigodisulfonic acid, Disodium salt
, Carmine Blue
, Food blue No. 2
, Sodium 3,3'-dioxo-[2,2'-biindolinylidene]-5,5'-disulphonate
, Sodium 3,3'-dioxo-[2,2'-biindolinylidene]-5,5'-disulfonate
, acid Blue 74
, 5,5'-Indigodisulphonic acid, Disodium salt
, FD and C Blue No. 2
, FD&C Blue #2
Indigo Carmine, Reagent, ACS, also known as 5,5'-indigodisulfonic acid sodium salt, is a pH indicator.
As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
Formula:
C₁₆H₈N₂Na₂O₈S₂ MW: 466.36 g/mol Melting Pt: > 300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005723 CAS Number: 860-22-0 EINECS: 212-728-8 Merck Index: 13,04969 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...