DL-Isoleucine ≥98% for biochemistry

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: H-DL-Ile-OH

I3238-5GM I3238-5KGBL
700007-338EA 57.7 USD
700007-338 700007-340
DL-Isoleucine ≥98% for biochemistry
DL-Isoleucine

DL-Isoleucine, also known as 2-Amino-3-methylpentanoic acid, is an alpha-amino acid and classified as a hydrophobic amino acid; it can be synthesized in a procedure starting from 2-bromobutane and diethylmalonate.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Melting Pt: 275
Density: 1.255 g/cm³ (–123.15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004268
CAS Number: 443-79-8
EINECS: 207-139-8
Merck Index: 13,05198

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR