1-Heptanol ≥98%, Reagent Grade

Supplier: Spectrum Chemical Mfg. Corp.
Warning

HE165-250ML HE165-20LT HE165-200LTB HE165-1LT HE165-2.5LT
700002-780EA 109.85 USD
700002-780 700002-778 700002-776 700002-772 700002-774
1-Heptanol ≥98%, Reagent Grade
1-Heptanol

1-Heptanol, Reagent

Formula: H₃C(CH₂)₅CH₂OH
Boiling Pt: 176 °C (1013 hPa)
Melting Pt: –35 °C
Density: 0.822
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00002986
CAS Number: 111-70-6
EINECS: 203-897-9
Merck Index: 13,04681

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR