3-Hydroxybenzoic acid

Supplier: Spectrum Chemical Mfg. Corp.
Warning

H2319-100GM H2319-25GM H2319-500GM
700006-496EA 117.4 USD
700006-496 700006-498 700006-500
3-Hydroxybenzoic acid
3-Hydroxybenzoic acid

m-Hydroxybenzoic Acid, also known as m-salicylic acid. It is a component of castoreum which is used in perfumery.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₇H₆O₃
Boiling Pt: 346 °C (1013 hPa)
Melting Pt: 199 °C
Density: 1.473 g/cm³ (20 °C)
Flash Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00002506
CAS Number: 99-06-9
EINECS: 202-726-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR