Guaiacol ≥99%, purified
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
2-Methoxyphenol
, 2-Hydroxyanisole
, 1-Hydroxy-2-methoxybenzene
GU101-500MLG
GU101-51002
GU101-100ML
700011-770EA
121.87
USD
CA11024-340
700011-770
700011-772
Guaiacol ≥99%, purified
Guaiacol
Guaiacol, Purified is a natural organic compound that is biosynthesized by several different organisms. It is a yellow pleasant smelling oil derived most commonly from wood creosote. The Purified Grade describes chemicals of good quality where there are no official standards and is normally limited to inorganic chemicals.
Formula:
C₇H₈O₂ MW: 124.14 g/mol Boiling Pt: 205 °C (1013 hPa) Melting Pt: 28…32 °C Density: 1.112 Flash Pt: 82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002185 CAS Number: 90-05-1 EINECS: 201-964-7 UN: 2810 ADR: 6.1,III Merck Index: 13,04568 |
Specification Test Results
Assay | 99% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...