L(-)-Glutathione (reduced form) ≥96%, reduced

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: L-Glutathione ReducedGlutathione Reduced(S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-mercapto-1-oxopropan-2-yl)amino)-5-oxopentanoic acidGSH

GL146-1GM GL146-5GM GL146-10KGBL GL146-100GM GL146-500GM GL146-25GM GL146-1KG GL146-250GM
700008-098EA 15673.91 USD
700008-098 700008-096 700008-102 700008-110 700008-100 700008-106 700008-104 700008-108
L(-)-Glutathione (reduced form) ≥96%, reduced
L(-)-Glutathione (reduced form)

L-Glutathione, Reduced can also be referred to as GSH and is an organic chemical that can be traced in plants and animals alike. It is an antioxidant that prevents damage from reactive oxygen species to cellular components that are important. It can be used to raise levels of plasma and liver GSH concentrations and is thought to have some positive effects on reducing cancer development.

Formula: C₁₀H₁₇N₃O₆S
MW: 307.33 g/mol
Melting Pt: 185…195 °C
Density: 1.475 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00065939
CAS Number: 70-18-8
EINECS: 200-725-4
Merck Index: 13,04488

Order Now

Specification Test Results

Assay 96% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR