Glutaric acid ≥98%, Reagent Grade for synthesis
Supplier: SPECTRUM CHEMICAL MFG CORP
|
Warning
|
Synonyms:
1,5-Pentanedioic acid
GL143-3KG
GL143-100GM
GL143-25GM
GL143-500GM
GL143-25KGBL
700005-932EA
2923.5
USD
700005-932
700005-926
700005-928
700005-934
700005-930
Glutaric acid ≥98%, Reagent Grade for synthesis
Glutaric acid
Glutaric Acid, Reagent, also known as 1,5-pentanediol, is an organic compound and is over 50% soluble in water. It is commonly used as a crosslinking agent. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.
Formula:
C₅H₈O₄ Boiling Pt: 302 °C (1013 hPa) Melting Pt: 98 °C Density: 1.43 g/cm³ (15 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004410 CAS Number: 110-94-1 EINECS: 203-817-2 Merck Index: 13,04486 |
Specification Test Results
Assay | 98% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...