Gold metal, powder 20 Mesh

Supplier: Spectrum Chemical Mfg. Corp.

G1201-250MG G1201-5GM G1201-1GM
700013-272EA 311.75 USD
700013-272 700013-274 700013-270
Gold metal, powder 20 Mesh
Formula: Au
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now

Specification Test Results

Particle size 20 Mesh


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR