Iron(II) sulfate hydrate, dried, powder USP
Supplier: Spectrum Chemical Mfg. Corp.
|
Warning
|
F1065-12KG
F1065-2.5KG
F1065-500GM
F1065-50KGBL
700001-552EA
1634.74
USD
700001-552
95033-246
95033-248
700001-554
Iron(II) sulfate hydrate, dried, powder USP
Iron (II) sulphate hydrate
Dried Ferrous Sulfate, Powder, USP is a mineral supplement.
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
FeSO₄·H₂O Melting Pt: ∼300 °C Density: 2.97 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149722 CAS Number: 13463-43-9 EINECS: 231-753-5 |
Specification Test Results
Assay | 86% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...