EDTA dipotassium salt dihydrate ≥99%, Reagent Grade

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Ethylenediaminetetraacetic acid dipotassium salt dihydrateEthylenedinitrilotetra-acetic acid dipotassium salt dihydrateEthylenedinitrilotetraacetic acid dipotassium saltTitriplex® Potassium saltEdetic acid dipotassium salt dihydrateEDTA dipotassium dihydrateDipotassium dihydrogen ethylenediaminetetraacetate dihydratePotassium 2,2'-((2-(bis(carboxymethyl)amino)ethyl)azanediyl)diacetate dihydrateEthylenediaminetetra-acetic acid dipotassium salt dihydrate

E1042-2.5KG E1042-50KGBL E1042-12KG E1042-100GM E1042-500GM
700007-276EA 138.81 USD
700007-276 700007-278 700007-284 700007-280 700007-282
EDTA dipotassium salt dihydrate ≥99%, Reagent Grade
EDTA dipotassium salt dihydrate

Dipotassium Dihydrogen Ethylenediaminetetraacetic Acid, Reagent

Formula: C₁₀H₁₄K₂N₂O₈·2H₂O
MW: 404.46 g/mol
Melting Pt: 272 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150036
CAS Number: 25102-12-9
EINECS: 217-895-0
Merck Index: 13,03546

Order Now

Specification Test Results

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR