Diethylene glycol monoethyl ether 99, reagent for synthesis
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
2-(2-Ethoxyethoxy)ethanol, Di(ethylene glycol) ethyl ether, Diethylene Glycol Monoethyl Ether
E1022-1LT
E1022-4LTGL
E1022-200LTB
E1022-20LTMT
700011-572EA
128.19
USD
700011-572
700011-578
700011-574
700011-576
Diethylene glycol monoethyl ether 99, reagent for synthesis
Diethylene glycol monoethyl ether
2-(2-Ethoxyethoxy)ethanol, Reagent is a clear, colorless, hygroscopic liquid that causes drying of skin by leaching fats, and is mildly irritating to the eyes. Its major use is an industrial solvent and its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.
Formula:
C₆H₁₄O₃ MW: 134.18 g/mol Boiling Pt: 197 Melting Pt: -80 Density: 0.991 Flash Pt: 93 Storage Temperature: Ambient |
MDL Number:
MFCD00002872 CAS Number: 111-90-0 EINECS: 203-919-7 REACH: 01-2119475105-42 Merck Index: 13,01809 |
Specification Test Results
Assay | 99% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...